Human Serum Albumin (HSA) Solution (25%), 50 ml
Catalog # GEMB800-120-050
Supplier: GEMINI BIO-PRODUCTS
New Product
Specifications
- Serum format:Frozen
- Sterility:Sterile
- Description:Human serum albumin solution
- Shipping conditions:Ships on dry ice
- Storage temperature:Store frozen at −20...−10 °C
- Pack type:Poly bottle
- Pk:50 mL
Specifications
About this item
This diagnostic-grade product is an aqueous solution containing 25% human albumin (w/v). It is prepared by cold-alcohol fractionation from pooled human plasma obtained from venous blood. The USP Pharmacopeia requires that all fractionators add sodium caprylate and sodium tryptophanate as a preservative.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.