Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Accessories for Data Logger
Accessories for Data Logger
IoT SIM card
Catalog # COMTLP105
Supplier:   COMET SYSTEM
Accessories for Data Logger
Catalog # COMTLP105
Supplier:  COMET SYSTEM
Accessories for Data Logger
Accessories for Data Logger
IoT SIM card
Price
22,61 €
Log in for availability
1 * 1 items
Product Details & Documents
A prepaid SIM card is an optional accessory for UxxxxM devices. It offers a long-term prepaid data connection to the mobile network, which allows sending values ​​from the device, for example, to the COMET Cloud.
  • The life of the SIM card is 2.5 years with a set sending interval of 5 minutes, 5 years with a sending interval of 10 minutes (500 MB)
  • Coverage includes more than 100 countries worldwide
The SIM card only supports data transfers, it does not support sending SMS messages.
Documents
Specifications
  • Description:
    IoT SIM card