Data Logger
Temperature recorder for four external Pt1000 probes
Catalog # COMTU0141
Supplier: COMET SYSTEM
Jump to:
Product Details & Documents
The datalogger is intended for recording temperatures from up to four external Pt1000 probes.
The recording is made to a non-volatile electronic memory. Data can be transferred to a personal computer at any time via the USB-C interface.
- The device contains a calibration sheet with declared metrological connections of standards, which is based on the requirements of the ČSN EN ISO/IEC 17025 standard
In case of exceeding the set limits, alarms are indicated on the device using LED, LCD and acoustic.
Specifications
- Description:Temperature recorder for four external Pt1000 probes