Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Data Logger
Data Logger
IoT wireless data logger of temperature, relative humidity with built-in 4G modem
Catalog # COMTU3120G
Supplier:   COMET SYSTEM
Data Logger
Catalog # COMTU3120G
Supplier:  COMET SYSTEM
Data Logger
Data Logger
IoT wireless data logger of temperature, relative humidity with built-in 4G modem
Price
506,78 €
Log in for availability
1 * 1 items
Product Details & Documents
The GSM data logger is intended for recording temperature and humidity. In case of exceeding the set limits, it allows sending alarm SMS and JSON messages via 4G data connection. Measured values ​​can be sent to the COMET Cloud Internet storage, which is a complete monitoring, alarming and analysis system.
  • The GSM recorder contains a calibration sheet with declared metrological connections of standards, which is based on the requirements of the ČSN EN ISO/IEC 17025 standard
It is also possible to set regular sending of JSON messages to the COMET Database program, the sending interval is adjustable. Alarms are also indicated locally on the device by LED, LCD and acoustically. The recording is made to a non-volatile electronic memory. The data can be transferred to a personal computer via the USB-C interface at any time with the supplied cable.
Specifications
  • Description:
    IoT wireless data logger of temperature, relative humidity with built-in 4G modem
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • IoT SIM card
    IoT SIM card
    COMTLP105
    Log in for availability
    1 * 1 items - 22,61 €