Data Logger
Product Details & Documents
The data logger is intended for temperature recording. The recording is carried out in a non-volatile electronic memory. The data can be transferred to a personal computer at any time via the USB interface, RS232, Ethernet or GSM modem for further processing.
- The device contains a calibration sheet with declared metrological connections of standards based on the requirements of the ČSN EN ISO/IEC 17025 standard
For communication with a PC, initialization and initial start-up, it is necessary to order a USB adapter or a COM adapter, or a start/stop magnet, from the Optional accessories, if you need to control the recording other than directly from the computer. The USB communication cable is not included in the package.
Specifications
- Description:Data logger for temperature recording with an internal sensor
Accessories
Product Information
- Catalog #AvailabilityPrice
- Wall holder for logger, gray with lock
Product Information
- Catalog #AvailabilityPrice
- Lithium battery 3.6 V/AA
Odporúčania budú personalizované na základe vašich nákupných preferencií len v prípade, že ste udelili súhlas aktiváciou možnosti "Vylepšiť môj nákupný zážitok"na stránke "Osobné údaje – Nastavenia ochrany osobných údajov“
V opačnom prípade dostanete všeobecné odporúčania.