Supelco® PESTANAL® Organic Reference Standard, Metolachlor
36163-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:51218-45-2
- Description:Metolachlor
- Seq:04
- Storage conditions:2...30 °C
- Hazard class description:H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055293
- M-Clarity quality level:MQ100
- Synonym(s):2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(-2-methoxy-1-methylethyl)acetamide
- Formula:C₁₅H₂₂ClNO₂
- Molecular weight:283,79
- Storage temperature:—
- Flash point:Not applicable
- GHS signal: GHS07
: P261 - P272 - P273 - P280 - P302 + P352 - P333 + P313