Supelco® PESTANAL® Organic Reference Standard, Metolachlor ESA Sodium Salt
34149-10MG
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:947601-85-6
- Description:Metolachlor ESA sodium salt
- Seq:03
- Storage conditions:2...30 °C
- Hazard class description:H317
- Shipping conditions:STD
- MDL No.:MFCD08702644
- M-Clarity quality level:MQ100
- Synonym(s):2-[(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)-amino]2-oxo-ethanesulfonic acid sodium salt
- Formula:C₁₅H₂₂NNaO₅S
- Molecular weight:351,39
- Storage temperature:—
- Flash point:Not applicable
- GHS signal: GHS07
: P280