Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, S-Metolachlor Metabolite CGA 37735
Supelco® PESTANAL® Organic Reference Standard, S-Metolachlor Metabolite CGA 37735
Catalog # 32645-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, S-Metolachlor Metabolite CGA 37735
Catalog # 32645-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    97055-05-5
  • Description:
    S-Metolachlor Metabolite CGA 37735
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD08442804
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-(2-Ethyl-6-methylphenyl)-2-hydroxyacetamid
  • Formula:
    C₁₁H₁₅NO₂
  • Molecular weight:
    193,24
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P301 + P312 - P302 + P352 - P305 + P351 + P338