Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Dicamba-D₃
  34233-10MG-R
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dicamba-D₃
  34233-10MG-R
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    349553-95-3
  • Description:
    Dicamba-d₃
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H318 - H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144104
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,6-Dichloro-2-(methoxy-d₃)benzoic acid
  • Formula:
    C₈D₃H₃Cl₂O₃
  • Molecular weight:
    224,06
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS05
 : P264 - P270 - P273 - P280 - P301 + P312 - P305 + P351 + P338