Anti-CD10 Mouse Monoclonal Antibody [Clone: C6/372 + 3F7B5]
Catalog # ANTIA253722-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Atriopeptidase
- Antigen symbol:CD10
- Clonality:Monoclonal
- Clone:C6/372 + 3F7B5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P30203
- Antigen synonyms:Enkephalinase|CD10|Common acute lymphocytic leukemia antigen|DKFZp686O16152|CALLA|Membrane metallo endopeptidase|EC 3.4.24.11|CD10 antigen|EPN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:90-130 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone C6/372: Recombinant human CD6 protein. Clone 3F7B5: Human rheumatoid synovial T cell line ST-1.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C6/372 + 3F7B5] antibody to CD10 for IF and IHC-P with samples derived from human.
- Validated Applications: IF, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: C6/372 + 3F7B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human