Order Entry
Slovakia
ContactUsLinkComponent
Anti-CD10 Rabbit Recombinant Antibody [clone: MME/8658R]
Anti-CD10 Rabbit Recombinant Antibody [clone: MME/8658R]
Catalog # ANTIA314577-100
Supplier:  ANTIBODIES.COM
New Product
Anti-CD10 Rabbit Recombinant Antibody [clone: MME/8658R]
Catalog # ANTIA314577-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Atriopeptidase
  • Antigen symbol:
    CD10
  • Clonality:
    Recombinant
  • Clone:
    MME/8658R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08473
  • Antigen synonyms:
    Enkephalinase|CD10|Common acute lymphocytic leukemia antigen|DKFZp686O16152|CALLA|Membrane metallo endopeptidase|EC 3.4.24.11|CD10 antigen|EPN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    100 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human CD10 protein fragment (aa 1-500). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MME/8658R] antibody to CD10 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: CD10
Clonality: Recombinant
Clone: MME/8658R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human