Anti-CD10 Rabbit Recombinant Antibody [clone: MME/8281R]
Catalog # ANTIA315959-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Atriopeptidase
- Antigen symbol:CD10
- Clonality:Recombinant
- Clone:MME/8281R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08473
- Antigen synonyms:Enkephalinase|CD10|Common acute lymphocytic leukemia antigen|DKFZp686O16152|CALLA|Membrane metallo endopeptidase|EC 3.4.24.11|CD10 antigen|EPN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:100 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human CD10 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MME/8281R] antibody to CD10 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD10
Clonality: Recombinant
Clone: MME/8281R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human