SLK Human shRNA Plasmid Kit (Locus ID 9748)
Catalog # ORIGTL320620
Supplier: ORIGENE TECHNOLOGIES
New Product
CAS Number:
Specifications
- Pk:1 KIT
- Description:SLK Human shRNA plasmid kit (Locus ID 9748)
- Details:E. coli selection: Chloramphenicol (34 µg/ml)
Mammalian cell selection: Puromycin
Format: Lentiviral plasmids - Vector details:pGFP-C-shLenti
Specifications
About this item
These shRNA constructs were designed against multiple splice variants at this gene locus.
- Mediates apoptosis and actin stress fiber dissolution
Delivery: Kit contains SLK - Human, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 9748) 5 µg purified plasmid DNA per construct, 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector.