Anti-Surface Glycoprotein Human Recombinant Monoclonal Antibody [clone: 2B7]
PRSI93-310
: PROSCI
- Antibody type:Primary
- Antigen name:surface glycoprotein
- Antigen symbol:S
- Clonality:Recombinant
- Clone:2B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG1
- Form:Liquid
- Gene ID:43740568
- Antigen synonyms:Spike protein|Surface Glycoprotein|SARS-CoV-2 (COVID-19) Spike S1 Antibody: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Modification:None
- Storage buffer:PBS, pH 7,4
- Storage temperature:24 months from manufacturing at <−20 °C; 1 month, 2...8 °C under sterile conditions
- Concentration:5,0 mg/ml (±10%)
- Shipping temperature:20
- Immunogen:Full length recombinant S Protein
- Purification:≥95%
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen: Surface Glycoprotein
Clonality: Recombinant Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Virus