Anti-Nucleocapsid Phosphoprotein Mouse Recombinant Monoclonal Antibody [clone: 4A6]
PRSI93-303
: PROSCI
- Antibody type:Primary
- Antigen name:nucleocapsid phosphoprotein
- Antigen symbol:N
- Clonality:Recombinant
- Clone:4A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Virus
- Form:Liquid
- Gene ID:43740575
- Antigen synonyms:N protein|Nucleocapsid Phosphoprotein|Nucleocapsid protein|SARS-CoV-2 (COVID-19) Nucleocapsid Antibody: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Modification:None
- Storage buffer:PBS, pH 7,4
- Storage temperature:24 months from manufacturing at <−20 °C; 1 month, 2...8 °C under sterile conditions
- Concentration:2,0 mg/ml (±10%)
- Shipping temperature:20
- Immunogen:Full length recombinant Nucleocapsid Protein
- Purification:≥95%
- Pack type:Vial
- Pk:0,1 mg
Type: Primary
Antigen: N
Clonality: Recombinant Monoclonal
Clone: 4A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus