Order Entry
Slovakia
ContactUsLinkComponent
 
Human Recombinant NGAL (from Cells)
  PRSI91-967
 :  PROSCI
 :  
Human Recombinant NGAL (from Cells)
  PRSI91-967
 :  PROSCI
 :  

 

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    25 kDa alpha-2-microglobulin-related subunit of MMP-9|Neutrophil gelatinase-associated lipocalin|Lipocalin-2|HNL|LCN2|Siderocalin LCN2|NGAL|Oncogene 24p3|p25
  • Protein/peptide name:
    NGAL
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21,6 kDa
  • Sequence:
    Gln21-Gly198
  • Formulation:
    Supplied as a 0,2 µm filtered solution of PBS, 50% glycerol, pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Used for biological assays

 

 

LCN2 is iron-trafficking protein involved in multiple processes such as apoptosis, innate immunity and renal development. LCN2 binds iron through association with 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin, and delivers or removes iron from the cell, depending on the context. LCN2 is involved in apoptosis due to interleukin-3 (IL3) deprivation: iron-loaded form increases intracellular iron concentration without promoting apoptosis, while iron-free form decreases intracellular iron levels, inducing expression of the proapoptotic protein BCL2L11/BIM, resulting in apoptosis. LCN2 is involved in innate immunity, possibly by sequestrating iron, leading to limit bacterial growth.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

 : For research use only.