Anti-Nucleoprotein Recombinant Antibody [clone: 4A1W0]
Catalog # PRSI11-087
Supplier: PROSCI
Specifications
- Antibody type:Primary
- Antigen name:nucleocapsid phosphoprotein
- Antigen symbol:Nucloprotein
- Clonality:Recombinant
- Clone:4A1W0
- Conjugation:Unconjugated
- ELISA:Yes
- Form:Liquid
- Gene ID:43740575
- Antigen synonyms:2019-nCoV N|2019nCoV N|2019-nCoV Nucleoprotein|2019-nCoV nucleocapsid protein.|SARS-CoV-2 NP|2019 peak N Protein|nucleocapsid protein [Severe acute respiratory syndrome coronavirus 2]|SARS-CoV-2 Nucleocapsid Protein|novel coronavirus Nucleocapsid Protein|novel coronavirus N Protein|2019-nCoV N Protein
- Modification:None
- Storage buffer:Supplied as a 0,2 µm filtered solution in PBS, pH 7,4
- Concentration:1 mg/ml
- Shipping temperature:20
- Immunogen:HEK293 derived SARS-COV-2 Nucleocapsid, amino acids Ser2-Ala419
- Purification:Affinity purified, endotoxin level: <1,0 EU/µg of the protein by LAL method
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Nucleoprotein
Clonality: Recombinant
Clone: 4A1W0
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG
Reactivity: Virus