Order Entry
Slovakia
ContactUsLinkComponent
Anti-TNF Recombinant Antibody [Clone: cA2 (Infliximab)]
Anti-TNF Recombinant Antibody [Clone: cA2 (Infliximab)]
Catalog # PRSI24-184
Supplier:  PROSCI
Anti-TNF Recombinant Antibody [Clone: cA2 (Infliximab)]
Catalog # PRSI24-184
Supplier:  PROSCI

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HGNC:HGNC:11892
  • Antigen symbol:
    TNF
  • Clonality:
    Recombinant
  • Clone:
    cA2 (Infliximab)
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Isotype:
    IgG kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    7124
  • Antigen synonyms:
    Tumor necrosis factor ligand superfamily member 2|TNFα|TNF-alpha|TNF-α|TNF-a|TNFSF|TNFa|Tumor necrosis factor|Cachectin
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Purified recombinant human TNF (rTNF)
  • Tested applications:
    In vivo assays
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: HGNC:HGNC:11892
Clonality: Recombinant
Clone: cA2 (Infliximab)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human