Anti-V5 Epitope Tag Recombinant Antibody [Clone: SV5-P-K]
Catalog # PRSI24-162
Supplier: PROSCI
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:SV5-P-K
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG4 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:V5 epitope tag|GKPIPNPLLGLDST epitope tag|PK tag|protein rev|Regulator of expression of viral proteins|PK-tag|GKPIPNPLLGLDST tag
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Pk (V5) Epitope Tag (GKPIPNPLLGLDST)
- Tested applications:RIA
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: V5 Epitope Tag
Clonality: Recombinant
Clone: SV5-P-K
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4 kappa
Reactivity: Paramyxovirus simian virus 5