Anti-CD52 Recombinant Antibody [Clone: Campath-1H]
PRSI24-124
: PROSCI
- Antibody type:Primary
- Antigen name:CD52 molecule
- Antigen symbol:CD52
- Clonality:Recombinant
- Clone:Campath-1H
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:1043
- Antigen synonyms:CDW52|Cambridge pathology 1 antigen|Epididymal secretory protein E5|Human epididymis-specific protein 5|YTH66.9HL|Alemtuzumab
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human lymphocytes
- Tested applications:Cytotoxicity assay, in vivo
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: CD52 molecule
Clonality: Recombinant
Clone: Campath-1H
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human;Rhesus monkey;Cynomolgus monkey