Anti-His tag Recombinant Antibody [Clone: 3D5]
Catalog # PRSI24-067
Supplier: PROSCI
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:3D5
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:polyhistidine tag
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Purified his tagged Fab fragment
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: His tag
Clonality: Recombinant
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: