Pesticide reference standard, Chlorpyrifos-methyl 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL11601000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:5598-13-0
- Description:Chlorpyrifos-methyl 10 µg/ml in cyclohexane
- Formula:C₇H₇Cl₃NO₃PS
- Description:Chlorpyrifos-methyl