Order Entry
Slovakia
ContactUsLinkComponent
Bromothymol blue sodium salt, water soluble indicator, Supelco®
Bromothymol blue sodium salt, water soluble indicator, Supelco®
Catalog # 1.01895.0010
Supplier:  Merck
CAS Number:  
Bromothymol blue sodium salt, water soluble indicator, Supelco®
Catalog # 1.01895.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Molar Mass:
    646.36 g/mol

Specification Test Results

About this item

 :
Melting Pt:> 200 °C
 :M-Clarity™ quality level = MQ200
 :646.37 g/mol
Storage Temperature:Ambient
EINECS:252-169-7
CAS Number:34722-90-2
Formula:C₂₇H₂₇Br₂NaO₅S
MDL Number:MFCD00077263