Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Trifluralin
Supelco® PESTANAL® Organic Reference Standard, Trifluralin
Catalog # 32061-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Trifluralin
Catalog # 32061-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1582-09-8
  • Description:
    Trifluralin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H317 - H351 - H410 - H302 - H317 - H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055241
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline
  • Molecular weight:
    335,28
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P273 - P280 - P202 - P273 - P280 - P301 + P312 - P302 + P352 - P308 + P313 - P301 + P312 - P302 + P352 - P308 + P313