Congo red, Sigma-Aldrich®
Catalog # SIALC6277-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:696.67 g/mol
Merck Index:12,02562
Melting Pt:> 360 °C
:M-Clarity™ quality level = MQ200
EINECS:209-358-4
:GHS08
Storage Temperature:Ambient
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028