Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Isovelleral
Isovelleral ≥99%
Isovelleral ≥99%
ENZOALX550356M001
:
ENZO LIFE SCIENCES
:
Isovelleral ≥99%
ENZOALX550356M001
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
Seq:
2070
Purity:
≥99%
:
:
232.3 g/mol
:
GHS07
:
Freezer
:
37841-91-1
:
C₁₅H₂₀O₂