Methotrexate
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
CAS Number:59-05-2
:MFCD00064370
Formula:C₂₀H₂₂N₈O₅
MDL Number:MFCD00150847
Molecular Weight:454.44 g/mol
Storage Temperature:Freezer
EINECS:200-413-8
UN:2811
ADR:6.1 III
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together