Order Entry
Slovakia
ContactUsLinkComponent
TSQ ≥98% (by HPLC), Ultrapure
TSQ ≥98% (by HPLC), Ultrapure
Catalog # ENZOENZ52153
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
TSQ ≥98% (by HPLC), Ultrapure
Catalog # ENZOENZ52153
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :328.38 g/mol
Storage Temperature:Freezer
CAS Number:109628-27-5
Formula:C₁₇H₁₆N₂O₃S