Sphingomyelin ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:85187-10-6
:MFCD00675919
Formula:C₄₁H₈₃N₂O₆P
MDL Number:MFCD00132361
:731.08 g/mol (stearoyl form)
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
Frequently Bought Together