Order Entry
Slovakia
ContactUsLinkComponent
BMOV ≥96% (by TLC)
BMOV ≥96% (by TLC)
Katalógové číslo ENZOBMLPR1310050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
BMOV ≥96% (by TLC)
Katalógové číslo ENZOBMLPR1310050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥96% (TLC)

Specification Test Results

About this item

 :
 :317.2 g/mol
Storage Temperature:Ambient
CAS Number:38213-69-3
Formula:C₁₂H₁₀O₇V