PP1 ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:172889-26-8
:
Formula:C₁₆H₁₉N₅
MDL Number:MFCD01076570
:281.4 g/mol
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Microspatulas, LaboPlast® / SteriPlast®
Supplier: BURKLE
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection