Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Endothall
Endothall ≥98%
Endothall ≥98%
Endothall ≥98%
Catalog # ENZOBMLPR1070100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Endothall ≥98%
Catalog # ENZOBMLPR1070100
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
186.163 g/mol
CAS Number:
145-73-3
Formula:
C₈H₁₀O₅
MDL Number:
MFCD00137362
Product Details