Order Entry
Slovakia
ContactUsLinkComponent
Prostaglandin E1 ≥99%
Prostaglandin E1 ≥99%
Catalog # ENZOBMLPG0060010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Prostaglandin E1 ≥99%
Catalog # ENZOBMLPG0060010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :354.487 g/mol
CAS Number:745-65-3
Formula:C₂₀H₃₄O₅
MDL Number:MFCD00077860