Z-Leu-Leu-Tyr-Ketoaldehyde
Catalog # ENZOBMLZW86550005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Protein/peptide name:Z-Leu-Leu-Tyr-Ketoaldehyde
Specifications
About this item
A ketoaldehyde. An excellent inhibitor of the chymotrypsin-like activity of the proteasome and serine and cysteine proteases in general.