Order Entry
Slovakia
ContactUsLinkComponent
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)
Irritant
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)
Catalog # ENZOALX106006M025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)
Catalog # ENZOALX106006M025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥97% (TLC)

Specification Test Results

About this item

 :
 :293.2 g/mol
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:220805-22-1
Formula:C₈H₁₈N₄O₂·2HCl·H₂O