


o-Tolidine, Supelco®
Catalog # 31659-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:212.29 g/mol
Merck Index:12,09651
Boiling Pt:∼361 °C (1013 hPa)
Melting Pt:129…131 °C
Density:∼1.234 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-358-0
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
:GHS09
CAS Number:119-93-7
Formula:[-C₆H₃(CH₃)-₄-NH₂]₂
MDL Number:MFCD00014773