0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standard, D-α-Phenylalanine 10 µg/ml in methanol
Organic reference standard, D-α-Phenylalanine 10 µg/ml in methanol
PROONE9305
:
PROMOCHEM
:
Organic reference standard, D-α-Phenylalanine 10 µg/ml in methanol
PROONE9305
:
PROMOCHEM
:
Pack type:
Glass bottle
Pk:
10 mL
CAS No.:
673-06-3
Description:
D-α-Phenylalanine 10 µg/ml in methanol
Formula:
C₉H₁₁NO₂
Description:
D-α-Phenylalanine