Anti-Nucleocapsid Phosphoprotein Rabbit Recombinant Monoclonal Antibody [clone: 1B1A9] (Biotinylated)
Catalog # PRSI24-043
Supplier: PROSCI
Specifications
- Antibody type:Primary
- Antigen name:Nucleocapsid phosphoprotein
- Antigen symbol:N
- Clone:1B1A9
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Form:Liquid
- Gene ID:1489678
- Antigen synonyms:2019-nCoV N|2019nCoV N|2019-nCoV Nucleoprotein|2019-nCoV nucleocapsid protein.|SARS-CoV-2 NP|2019 peak N Protein|nucleocapsid protein [Severe acute respiratory syndrome coronavirus 2]|SARS-CoV-2 Nucleocapsid Protein|novel coronavirus Nucleocapsid Protein|novel coronavirus N Protein|2019-nCoV N Protein
- Modification:None
- Storage buffer:Supplied as a 0,2 μm filtered solution in PBS, pH 7,4 containing SARS-CoV-2 (2019-nCoV) nucleocapsid antibody at a concentration of 1 mg/ml.
- Storage temperature:This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at −20...−80 °C. Preservative-Free.Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:20
- Immunogen:Hek293 derived SARS-COV-2 Nucleocapsid Ser2-Ala419 Accession #YP_009724397.2.
- Purification:Affinity purification 1 EU/µg of the rotein by LAL method.
- Size:0,1 mg
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: N
Clonality: Recombinant Monoclonal
Clone: 1B1A9
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus