Order Entry
Slovakia
ContactUsLinkComponent
Ethanol 70% ≥70% (v/v)
Ethanol 70% ≥70% (v/v)
Catalog # 93003.1006
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ethanol 70% ≥70% (v/v)
Catalog # 93003.1006
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Seq:
    5055
  • Environmentally Preferable:
  • Pk Info:
    6 x 1 L

Specifications

Specification Test Results

  • Assay (Ethanol) (V/V):
    Min. 69,9 %
  • Appearance:
    Clear colourless liquid
  • Relative density:
    0.880 - 0.890

Specification Test Results

About this item

Synonyms: Ethyl alcohol
 :
 :46.07 g/mol
EINECS:200-578-6
UN:1170
 :GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568
Boiling Pt:78.3
Melting Pt:-117
Flash Pt:22 °C
ADR:3 II
Merck Index:12,03806
 :Filtered through a 0.2 µm filter.