
Adiponitrile, Sigma-Aldrich®
Catalog # 8.00092.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:50 g/l (20 °C)
- Melting Point:2 °C
- Molar Mass:108.14 g/mol
- Boiling Point:295 °C
- Vapor Pressure:0.003 hPa (20 °C)
- Flash Point:93 °C (decomposition)
- Refractive Index:1.438 (20 °C, 589 nm)
- Explosion Limit:7 - 14 %(V)
- Density:0.96 g/cm3 (20 °C)
- Ignition Point:460 °C
Specification Test Results
About this item
:
:108.14 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:2 °C
Density:0.965 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-896-3
UN:2205
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS06
CAS Number:111-69-3
Formula:C₆H₈N₂
MDL Number:MFCD00001975