Supelco® PESTANAL® Organic Reference Standard, DNOC
Catalog # 45464-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:534-52-1
- Description:DNOC
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 + H330 - H315 - H317 - H318 - H341 - H410
- Shipping conditions:STD
- MDL No.:MFCD00007105
- M-Clarity quality level:MQ100
- Synonym(s):2-Methyl-4,6-dinitrophenol; 3,5-Dinitro-2-hydroxytoluene; 4,6-Dinitro-2-methylphenol; 4,6-Dinitro-o-cresol; DNOC
- Molecular weight:198,13
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS05, GHS09
Caution: P201 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310 - P305 + P351 + P338