
Cinnamaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-55-2
:
Formula:C₉H₈O
MDL Number:MFCD00007000
:132.16 g/mol
Merck Index:12,02356
Boiling Pt:251 °C (1013 hPa)
Melting Pt:–8 °C
Density:1.112 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:138 °C
EINECS:203-213-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together