Sodium nitroprusside dihydrate ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: HONEYWELL
Certificates
About this item
CAS Number:13755-38-9
:
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:238-373-9
UN:3288
ADR:6.1,III
:297.95 g/mol
:GHS06
Specification Test Results
- Assay:99.0 - 101.0 %
- Water insoluble matter:Max. 0.01 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 50 ppm
- Copper (Cu):Max. 5 ppm
- Potassium (K):Max. 3000 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 50 ppm
- Chloride (Cl):Max. 100 ppm
- Sulfate (SO₄):Max. 100 ppm
- Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻):Max. 200 ppm
- Hexacyanoferrate(III) ([Fe(CN)₆]₃⁻):Max. 100 ppm
Specification Test Results
Frequently Bought Together