Supelco® PESTANAL® Organic Reference Standard, Amitrol
Catalog # 45324-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:61-82-5
- Description:Amitrol
- Storage conditions:–10…–25 °C
- Hazard class description:H361d - H373 - H411
- Shipping conditions:STD
- MDL No.:MFCD00005230
- M-Clarity quality level:MQ100
- Synonym(s):3-Amino-1,2,4-triazole; 1,2,4-Triazol-3-amine; 3-AT; Amitrol
- Melting point:150...153 °C (lit.)
- Molecular weight:84,08
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P260 - P273 - P280 - P308 + P313 - P391