Order Entry
Slovakia
ContactUsLinkComponent
Dithizone, Sigma-Aldrich®
Irritant
Dithizone, Sigma-Aldrich®
Catalog # SIAL43820-10G
Supplier:  Merck
CAS Number:  
Irritant
Dithizone, Sigma-Aldrich®
Catalog # SIAL43820-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :256.33 g/mol
Merck Index:13,03413
Boiling Pt:406 °C (1013 hPa)
Melting Pt:168 °C
EINECS:200-454-1
 :GHS07
CAS Number:60-10-6
Formula:C₆H₅NHNHCSN=NC₆H₅
MDL Number:MFCD00003025