Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Eucalyptol
Supelco® Organic Reference Standard, Eucalyptol
Catalog # 29210-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Eucalyptol
Catalog # 29210-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    470-82-6
  • Description:
    Eucalyptol
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H226 - H317
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00167977
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane; 1,8-Cineole; 1,8-Epoxy-p-menthane
  • Melting point:
    1...2 °C (lit.)
  • Molecular weight:
    154,25
  • Density:
    0,921 g/ml at 25 °C (lit.)
  • Flash point:
    52 °C - closed cup
  • Boiling point:
    176...177 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02
Caution: P210 - P280 - P302 + P352