Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Acetone

Flammable
Irritant
   Sustainable Options Available
Supplier:   SCHARLAB
Acetone
Supplier:  SCHARLAB
   Sustainable Options Available
Certificates
Acetone

2 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SCARAC03081000
  • SCARAC03082500
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1.000 mL
    Log in for availability
    Log in for availability
    1 * 1.000 mL
    1 * 1.000 mL

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SCARAC03081000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SCARAC03082500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health