Supelco® Organic Reference Standard, Dibenzothiophene
Catalog # 45776-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:132-65-0
- Description:Dibenzothiophene
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00004969
- M-Clarity quality level:MQ100
- Synonym(s):DBT; Diphenylene sulfide
- Melting point:97 - 100 °C (lit.)
- Molecular weight:184,26
- Flash point:170 °C
- Boiling point:332 - 333 °C (lit.)
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P301 + P312 + P330