Order Entry
Slovakia
ContactUsLinkComponent
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium(0)
Catalog # ABCRAB107083.0025
Supplier:  abcr
CAS Number:  
Tetrakis(triphenylphosphine)palladium(0)
Catalog # ABCRAB107083.0025
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :1155.58 g/mol
Melting Pt:103…107 °C
Density:1 g/cm³ (20 °C)
EINECS:238-086-9
CAS Number:14221-01-3
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012