Aspartame
EHERC10304940
:
- Pk:250 mg
- CAS No.:22839-47-0
- MDL No.:MFCD00002724
- Protein synonyms:H-Asp-Phe-OMe|(3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid|L-AspartyL-L-phenylalanine methyl ester|N-L-α-Aspartyl-L-phenylalanine 1-Methyl Ester
- Protein/peptide name:Aspartame
- Molecular weight:294,30 g/mol
- Molecular formula:C₁₄H₁₈N₂O₅